EMAIL THIS PAGE TO A FRIEND
495123 Sigma-Aldrich

Barium molybdate

99.9% trace metals basis

Purchase

Properties

Related Categories Barium Salts, Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Materials Science,
InChI Key   RCMWGBKVFBTLCW-UHFFFAOYSA-N
assay   99.9% trace metals basis
form   powder
density   4.65 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 1564 6.1 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?