• USA Home
  • 496049 - (S)-(+)-4-(1H-Indol-3-ylmethyl)-2-oxazolidinone

EMAIL THIS PAGE TO A FRIEND
496049 Sigma-Aldrich

(S)-(+)-4-(1H-Indol-3-ylmethyl)-2-oxazolidinone

98%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Auxiliaries, Oxazolidinone Derivatives
assay   98%
optical activity   [α]21/D +9°, c = 1 in methanol
mp   157-160 °C (lit.)
SMILES string   O=C1N[C@H](CO1)Cc2c[nH]c3ccccc23
InChI   1S/C12H12N2O2/c15-12-14-9(7-16-12)5-8-6-13-11-4-2-1-3-10(8)11/h1-4,6,9,13H,5,7H2,(H,14,15)/t9-/m0/s1
InChI key   LGXHODFXCOIGGJ-VIFPVBQESA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?