• USA Home
  • 496995 - 4-O-Benzyl-D-galactal

EMAIL THIS PAGE TO A FRIEND
496995 Sigma-Aldrich

4-O-Benzyl-D-galactal

97%

  • CAS Number 157380-76-2

  • Empirical Formula (Hill Notation) C13H16O4

  • Molecular Weight 236.26

  •  PubChem Substance ID 24873096

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
assay   97%
optical activity   [α]20/D −22°, c = 1 in chloroform
mp   101-103 °C (lit.)
SMILES string   OC[C@H]1OC=C[C@@H](O)[C@H]1OCc2ccccc2
InChI   1S/C13H16O4/c14-8-12-13(11(15)6-7-16-12)17-9-10-4-2-1-3-5-10/h1-7,11-15H,8-9H2/t11-,12-,13-/m1/s1
InChI key   OIWQZGBSQDJVJN-JHJVBQTASA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?