• USA Home
  • 497037 - 4,4′′-Dimethyl-4′-(4-methylphenyl)-2,2′:6′,2′′-terpyridine

EMAIL THIS PAGE TO A FRIEND
497037 Sigma-Aldrich

4,4′′-Dimethyl-4′-(4-methylphenyl)-2,2′:6′,2′′-terpyridine

98%

  • CAS Number 302912-20-5

  • Empirical Formula (Hill Notation) C24H21N3

  • Molecular Weight 351.44

  •  MDL number MFCD01863612

Purchase

Properties

assay   98%
mp   244-247 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?