• USA Home
  • 497436 - Dichloro[rac-ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]titanium(IV)

EMAIL THIS PAGE TO A FRIEND
497436 Sigma-Aldrich

Dichloro[rac-ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]titanium(IV)

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Titanium Catalysts More...
reaction suitability   reagent type: catalyst
mp   256-261 °C (dec.) (lit.)
SMILES string   Cl[Ti]Cl.[CH]1[CH][C](CC[C]2[CH][CH][C]3CCCC[C]23)[C]4CCCC[C]14
InChI   1S/C20H24.2ClH.Ti/c1-3-7-19-15(5-1)9-11-17(19)13-14-18-12-10-16-6-2-4-8-20(16)18;;;/h9-12H,1-8,13-14H2;2*1H;/q;;;+2/p-2
InChI key   JCEDTJKHTKLRJD-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 1759 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?