• USA Home
  • 497460 - [(R,R)-Ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]dimethyltitanium(IV)

EMAIL THIS PAGE TO A FRIEND
497460 Sigma-Aldrich

[(R,R)-Ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]dimethyltitanium(IV)

  • CAS Number 160335-85-3

  • Empirical Formula (Hill Notation) C22H30Ti

  • Molecular Weight 342.34

  •  MDL number MFCD01863619

Purchase

Properties

optical activity   [α]21/D −3006°, c = 0.07 in methylene chloride
mp   76-80 °C (dec.) (lit.)
SMILES string   C[Ti]1(C)[C@]4(CC[C@]12C=CC3=C2CCCC3)C=CC5=C4CCCC5
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?