• USA Home
  • 498122 - 3,4-Di-O-acetyl-6-O-(triisopropylsilyl)-D-galactal

EMAIL THIS PAGE TO A FRIEND
498122 Aldrich

3,4-Di-O-acetyl-6-O-(triisopropylsilyl)-D-galactal

97%

Purchase

Properties

InChI Key   QKFMTAHXVXFZEZ-GUDVDZBRSA-N
assay   97%
optical activity   [α]22/D −24°, c = 3 in chloroform
mp   25-29 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?