• USA Home
  • 498130 - 3,4-Di-O-acetyl-6-O-(tert-butyldiphenylsilyl)-D-galactal

EMAIL THIS PAGE TO A FRIEND
498130 Aldrich

3,4-Di-O-acetyl-6-O-(tert-butyldiphenylsilyl)-D-galactal

97%

Purchase

Properties

InChI Key   HGWFRQHLOUXHGQ-UBFVSLLYSA-N
assay   97%
optical activity   [α]22/D −4°, c = 1.1 in chloroform
contains   3-5 wt. % ethyl acetate, added to induce mobility of syrup
refractive index   n20/D 1.526 (lit.)
Safety & Documentation

Safety Information

Flash Point(F) 
150 °F
Flash Point(C) 
66 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?