• USA Home
  • 498149 - 4-O-Acetyl-3,6-di-O-(triisopropylsilyl)-D-galactal

EMAIL THIS PAGE TO A FRIEND
498149 Aldrich

4-O-Acetyl-3,6-di-O-(triisopropylsilyl)-D-galactal

97%

Purchase

Properties

InChI Key   QRUQNCIVNNLVGC-TWJOJJKGSA-N
assay   97%
optical activity   [α]23/D −39°, c = 1 in chloroform
refractive index   n20/D 1.475 (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?