• USA Home
  • 498416 - 3,4-Dimethylphenylzinc iodide solution

EMAIL THIS PAGE TO A FRIEND
498416 Sigma-Aldrich

3,4-Dimethylphenylzinc iodide solution

0.5 M in THF

Purchase

Properties

Related Categories Aryl, Chemical Synthesis, Organometallic Reagents, Organozinc Halides, Rieke and Organozinc Reagents More...
InChI Key   XWAXMHZTAPXNAV-UHFFFAOYSA-M
concentration   0.5 M in THF
bp   65 °C
density   1.015 g/mL at 25 °C

Description

Packaging

50 mL in Sure/Seal™

Legal Information

Product of Rieke® Metals, Inc. ®Registerd trademark of Rieke Metals, Inc.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?