• USA Home
  • 498432 - 3,5-Dimethyl-1-adamantylzinc bromide solution

EMAIL THIS PAGE TO A FRIEND
498432 Sigma-Aldrich

3,5-Dimethyl-1-adamantylzinc bromide solution

0.5 M in THF

Purchase

Properties

concentration   0.5 M in THF
bp   65 °C

Description

Legal Information

Product of Rieke® Metals, Inc.

Rieke is a registered trademark of Rieke Metals, Inc.

Safety & Documentation

Safety Information

RIDADR 
UN 2056 3/PG 2
WGK Germany 
1
Flash Point(F) 
1 °F
Flash Point(C) 
-17 °C
Protocols & Articles

Articles

Rieke® Organozincs

Sigma-Aldrich, in collaboration with Rieke®, is proud to offer a variety of organozinc reagents in research-scale quantities.
Aldrich ChemFiles 2006, 6.3, 10.
Keywords: Applications, Catalysis, Chemfiles, Cross couplings

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?