• USA Home
  • 498629 - 5-Fluoro-2-methylphenylzinc iodide solution

EMAIL THIS PAGE TO A FRIEND
498629 Sigma-Aldrich

5-Fluoro-2-methylphenylzinc iodide solution

0.5 M in THF

Purchase

Properties

Related Categories Aryl, Chemical Synthesis, Organometallic Reagents, Organozinc Halides, Rieke and Organozinc Reagents More...
concentration   0.5 M in THF
bp   65 °C
density   1.012 g/mL at 25 °C
storage temp.   2-8°C
SMILES string   Cc1ccc(F)cc1[Zn]I
InChI   1S/C7H6F.HI.Zn/c1-6-2-4-7(8)5-3-6;;/h2,4-5H,1H3;1H;/q;;+1/p-1
InChI key   PRGNEWAVDWRJKY-UHFFFAOYSA-M

Description

Packaging

50 mL in Sure/Seal™

Legal Information

Product of Rieke® Metals, Inc.

Rieke is a registered trademark of Rieke Metals, Inc.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?