EMAIL THIS PAGE TO A FRIEND
49874 Aldrich

Boc-Met(O2)-OH

≥96.0% (TLC)

Synonym: Boc-L-methionine sulfone

  • CAS Number 60280-45-7

  • Empirical Formula (Hill Notation) C10H19NO6S

  • Molecular Weight 281.33

  •  MDL number MFCD00065587

  •  PubChem Substance ID 57651227

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Methionine, Peptide Chemistry | Peptide Synthesis More...
InChI Key   CPGDRHNBSOQFMF-ZETCQYMHSA-N
assay   ≥96.0% (TLC)
optical activity   [α]20/D −13.5±1°, c = 1% in DMF
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?