• USA Home
  • 498939 - 2-Methylphenylzinc iodide solution

EMAIL THIS PAGE TO A FRIEND
498939 Sigma-Aldrich

2-Methylphenylzinc iodide solution

0.5 M in THF

Purchase

Properties

concentration   0.5 M in THF
density   0.987 g/mL at 25 °C

Description

Legal Information

Product of Rieke® Metals, Inc.

Rieke is a registered trademark of Rieke Metals, Inc.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?