• USA Home
  • 499927 - 5-Acetyl-2-thienylboronic acid

EMAIL THIS PAGE TO A FRIEND
499927 Sigma-Aldrich

5-Acetyl-2-thienylboronic acid

Synonym: (2-Acetylthien-5-yl)boronic acid, (5-Acetylthien-2-yl)boronic acid, 5-Acetyl-2-thiopheneboronic acid, 5-Acetylthiophen-2-ylboronic acid, 5-Acetylthiophene-2-boronic acid

Purchase

Properties

Related Categories Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Boronic Acids, Organometallic Reagents More...
mp   133-138 °C (lit.)
SMILES string   CC(=O)c1ccc(s1)B(O)O
InChI   1S/C6H7BO3S/c1-4(8)5-2-3-6(11-5)7(9)10/h2-3,9-10H,1H3
InChI key   DCNMATSPQKWETQ-UHFFFAOYSA-N

Description

Application

Reactant for:
Bioreduction by alcohol dehydrogenase
Preparation of highly fluorescent photochromic dyes by photocyclization reactions
Suzuki-Miyaura cross-couplings

Packaging

5, 25 g in glass bottle

Other Notes

Contains varying amounts anhydride

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Arylboronic Acids

We are pleased to present its complete selection of arylboronic acids, the building blocks of the Suzuki coupling experiment. Most arylboronic acids readily undergo dehydration reactions to give a cy...
ChemFiles 2001, 1.1, 5.
Keywords: Building blocks, Coupling reactions, Dehydration reaction, Suzuki coupling

Suzuki–Miyaura Coupling Reagents

This brochure contains a comprehensive selection of boronic acids, boronic acid esters, diboron esters, and transition-metal catalysts useful for the Suzuki–Miyaura coupling reaction. New offerings a...
ChemFiles Volume 4 Article 2
Keywords: Coupling reactions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?