• USA Home
  • 499951 - 4-(Dibenzofuranyl)boronic acid

EMAIL THIS PAGE TO A FRIEND
499951 Sigma-Aldrich

4-(Dibenzofuranyl)boronic acid

≥95.0%

Synonym: 4-Dibenzofuranboronic acid

  • CAS Number 100124-06-9

  • Empirical Formula (Hill Notation) C12H9BO3

  • Molecular Weight 212.01

  •  MDL number MFCD00092336

  •  PubChem Substance ID 24873313

  •  NACRES NA.22

Purchase

Properties

Related Categories Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Boronic Acids, Organometallic Reagents More...
Quality Level   100
assay   ≥95.0%
mp   286-291 °C (lit.)
SMILES string   OB(O)c1cccc2c3ccccc3oc12
InChI   1S/C12H9BO3/c14-13(15)10-6-3-5-9-8-4-1-2-7-11(8)16-12(9)10/h1-7,14-15H
InChI key   ZXHUJRZYLRVVNP-UHFFFAOYSA-N

Description

Packaging

5 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Arylboronic Acids

We are pleased to present its complete selection of arylboronic acids, the building blocks of the Suzuki coupling experiment. Most arylboronic acids readily undergo dehydration reactions to give a cy...
ChemFiles 2001, 1.1, 5.
Keywords: Building blocks, Coupling reactions, Dehydration reaction, Suzuki coupling

Suzuki–Miyaura Coupling Reagents

This brochure contains a comprehensive selection of boronic acids, boronic acid esters, diboron esters, and transition-metal catalysts useful for the Suzuki–Miyaura coupling reaction. New offerings a...
ChemFiles Volume 4 Article 2
Keywords: Coupling reactions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?