EMAIL THIS PAGE TO A FRIEND
510815 Aldrich

Diphenylpropynone

Synonym: 2-Benzoyl-1-phenylacetylene

Purchase

Properties

bp   136-139 °C/3 mmHg(lit.)
mp   41-46 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UK5122000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

We list over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of these are termin...
Keywords: Building blocks, Coupling reactions, Cross couplings

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?