• USA Home
  • 51195 - L-Gulonic acid γ-lactone

EMAIL THIS PAGE TO A FRIEND
51195 Sigma-Aldrich

L-Gulonic acid γ-lactone

purum, ≥98.0% (T)

Synonym: L(+)-Gulono-1,4-lactone

  • CAS Number 1128-23-0

  • Empirical Formula (Hill Notation) C6H10O6

  • Molecular Weight 178.14

  •  Beilstein/REAXYS Number 83002

  •  MDL number MFCD00064331

  •  PubChem Substance ID 24873594

Purchase

Properties

grade   purum
assay   ≥98.0% (T)
optical activity   [α]20/D +55±1°, c = 10% in H2O
mp   183-187 °C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

310301 L-Gulonic acid γ-lactone, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?