• USA Home
  • 512575 - Bis(butylcyclopentadienyl)tungsten(IV) dichloride

EMAIL THIS PAGE TO A FRIEND
512575 Sigma-Aldrich

Bis(butylcyclopentadienyl)tungsten(IV) dichloride

97%

  • CAS Number 87695-18-9

  • Empirical Formula (Hill Notation) C18H26Cl2W

  • Molecular Weight 497.15

  •  MDL number MFCD02093657

  •  PubChem Substance ID 24873643

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Tungsten Catalysts More...
assay   97%
mp   107-109 °C (dec.) (lit.)
SMILES string   CCCC[C]=12C3C4=C5C=1[W]23456789(Cl)(Cl)C%10C6=C7C8=[C]9%10CCCC
InChI   1S/2C9H13.2ClH.W/c2*1-2-3-6-9-7-4-5-8-9;;;/h2*4-5,7-8H,2-3,6H2,1H3;2*1H;/q;;;;+2/p-2
InChI key   UQKICGNQFKTSSH-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?