• USA Home
  • 512664 - N-(2,3-Epoxypropyl)phthalimide

EMAIL THIS PAGE TO A FRIEND
512664 Aldrich

N-(2,3-Epoxypropyl)phthalimide

90%

Synonym: N-Glycidylphthalimide

  • CAS Number 5455-98-1

  • Empirical Formula (Hill Notation) C11H9NO3

  • Molecular Weight 203.19

  •  Beilstein/REAXYS Number 171277

  •  EC Number 226-710-2

  •  MDL number MFCD00005896

Purchase

Properties

InChI Key   DUILGEYLVHGSEE-UHFFFAOYSA-N
assay   90%
availability   available only in EU
mp   92-98 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
TI4950000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?