• USA Home
  • 513601 - Ethyl 3,3-bis(tert-amylperoxy)butyrate solution

EMAIL THIS PAGE TO A FRIEND
513601 Sigma-Aldrich

Ethyl 3,3-bis(tert-amylperoxy)butyrate solution

75 wt. % in odorless mineral spirits

Synonym: Luperox® 533M75

  • CAS Number 67567-23-1

  • Linear Formula CH3C[OOC(CH3)2C2H5]2CH2CO2C2H5

  • Molecular Weight 320.42

  •  MDL number MFCD01863713

Purchase

Properties

concentration   75 wt. % in odorless mineral spirits
refractive index   n20/D 1.4285
density   0.894 g/mL at 25 °C

Description

Application

Polymerization initiator

Legal Information

Product of Arkema Inc.

Luperox is a registered trademark of Arkema Inc.

Safety & Documentation

Safety Information

RIDADR 
UN 3105 5.2
WGK Germany 
3
Flash Point(F) 
115 °F
Flash Point(C) 
46 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?