• USA Home
  • 51360 - Harmalol hydrochloride dihydrate

EMAIL THIS PAGE TO A FRIEND
51360 Sigma-Aldrich

Harmalol hydrochloride dihydrate

≥98.0% (AT)

Synonym: 4,9-Dihydro-1-methyl-3H-pyrido[3,4-b]indol-7-ol hydrochloride

  • CAS Number 6028-00-8

  • Empirical Formula (Hill Notation) C12H12N2O · HCl · 2H2O

  • Molecular Weight 272.73

  •  Beilstein/REAXYS Number 3729233

  •  MDL number MFCD00150053

  •  PubChem Substance ID 24873701

Purchase

Properties

assay   ≥98.0% (AT)
drug control   stupéfiantregulated under CDSA - not available from Sigma-Aldrich Canada
mp   265-268 °C (dec.)

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?