• USA Home
  • 513679 - tert-Butyl peracetate (Luperox® 7M50) solution

EMAIL THIS PAGE TO A FRIEND
513679 Sigma-Aldrich

tert-Butyl peracetate (Luperox® 7M50) solution

50 wt. % in aliphatic hydrocarbons

Synonym: tert-Butyl peracetate solution

  • CAS Number 107-71-1

  • Linear Formula CH3CO3C(CH3)3

  • Molecular Weight 132.16

  •  MDL number MFCD00048240

Purchase

Properties

InChI Key   SWAXTRYEYUTSAP-UHFFFAOYSA-N
concentration   50 wt. % in aliphatic hydrocarbons
refractive index   n20/D 1.413
density   0.83 g/mL at 25 °C

Description

Application

Polymerization initiator.

Legal Information

Product of Arkema Inc.

Luperox is a registered trademark of Arkema Inc.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 3103A7 5.2
WGK Germany 
3
Flash Point(F) 
98.6 °F
Flash Point(C) 
37 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

388076 tert-Butyl peracetate solution, 50 wt. % in odorless mineral spirits

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?