• USA Home
  • 513792 - 3,6-Di-O-(triisopropylsilyl)-D-glucal

EMAIL THIS PAGE TO A FRIEND
513792 Sigma-Aldrich

3,6-Di-O-(triisopropylsilyl)-D-glucal

97%

  • Empirical Formula (Hill Notation) C24H50O4Si2

  • Molecular Weight 458.82

  •  MDL number MFCD01863720

  •  PubChem Substance ID 24873713

  •  NACRES NA.22

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
assay   97%
optical activity   [α]25/D −22°, c = 1 in chloroform
refractive index   n20/D 1.48 (lit.)
density   0.965 g/mL at 25 °C (lit.)
storage temp.   −20°C
SMILES string   CC(C)[Si](OC[C@H]1OC=C[C@@H](O[Si](C(C)C)(C(C)C)C(C)C)[C@@H]1O)(C(C)C)C(C)C
InChI   1S/C24H50O4Si2/c1-16(2)29(17(3)4,18(5)6)27-15-23-24(25)22(13-14-26-23)28-30(19(7)8,20(9)10)21(11)12/h13-14,16-25H,15H2,1-12H3/t22-,23-,24+/m1/s1
InChI key   YDOYFAHLHUEGEN-SMIHKQSGSA-N

Description

Packaging

500 mg in amber glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?