EMAIL THIS PAGE TO A FRIEND
514411 Sigma-Aldrich

3-Chlorophenetole

97%

Purchase

Properties

Related Categories Building Blocks, C8, Chemical Synthesis, Ethers, Organic Building Blocks,
InChI Key   HNMYSCABMYNGTP-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.525 (lit.)
bp   86.5 °C/12 mmHg (lit.)
density   1.12 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
197.6 °F
Flash Point(C) 
92 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?