• USA Home
  • 514454 - 2,2-Dibutyl-1,3-propanediol

EMAIL THIS PAGE TO A FRIEND
514454 Sigma-Aldrich

2,2-Dibutyl-1,3-propanediol

97%

  • CAS Number 24765-57-9

  • Linear Formula [CH3(CH2)3]2C(CH2OH)2

  • Molecular Weight 188.31

  •  MDL number MFCD01863725

  •  PubChem Substance ID 24873760

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
Quality Level   100
assay   97%
mp   41-43 °C (lit.)
SMILES string   CCCCC(CO)(CO)CCCC
InChI   1S/C11H24O2/c1-3-5-7-11(9-12,10-13)8-6-4-2/h12-13H,3-10H2,1-2H3
InChI key   OJMJOSRCBAXSAQ-UHFFFAOYSA-N

Description

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
235.4 °F - closed cup
Flash Point(C) 
113 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?