• USA Home
  • 514810 - Bis[[4-[(vinyloxy)methyl]cyclohexyl]methyl] terephthalate

EMAIL THIS PAGE TO A FRIEND
514810 Sigma-Aldrich

Bis[[4-[(vinyloxy)methyl]cyclohexyl]methyl] terephthalate

90%

Synonym: VEctomer® 4051 vinyl ether

  • CAS Number 209072-72-0

  • Linear Formula C6H4-1,4-[CO2CH2C6H10CH2OCH=CH2]2

  • Molecular Weight 470.60

  •  MDL number MFCD01863727

Purchase

Properties

assay   90%
SMILES string   C=COCC1CCC(CC1)COC(=O)c2ccc(cc2)C(=O)OCC3CCC(CC3)COC=C

Description

Legal Information

Product of Morflex, Inc. ®Registered trademark of Morflex, Inc.

VEctomer is a registered trademark of Vertellus Specialties, Inc.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
334.4 °F
Flash Point(C) 
168 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?