• USA Home
  • 514969 - γ-Methyl-α-(trifluoromethyl)-γ-valerolactone

EMAIL THIS PAGE TO A FRIEND
514969 Sigma-Aldrich

γ-Methyl-α-(trifluoromethyl)-γ-valerolactone

96%

Synonym: Dihydro-5,5-dimethyl-3-(trifluoromethyl)-2(3H)-furanone

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Lactones, Organic Building Blocks More...
assay   96%
refractive index   n20/D 1.389 (lit.)
bp   216 °C (lit.)
density   1.25 g/mL at 25 °C (lit.)
SMILES string   CC1(C)CC(C(=O)O1)C(F)(F)F
InChI   1S/C7H9F3O2/c1-6(2)3-4(5(11)12-6)7(8,9)10/h4H,3H2,1-2H3
InChI key   NSQKIHBAKTVICL-UHFFFAOYSA-N

Description

Packaging

1 mL in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?