EMAIL THIS PAGE TO A FRIEND
515043 Sigma-Aldrich

Toluene-d8

99.6 atom % D

Purchase

Properties

Related Categories NMR Solvents, Solvent by Type, Solvent by Application, Solvents, Spectroscopy Solvents (IR, NMR, UV/Vis),
InChI Key   YXFVVABEGXRONW-JGUCLWPXSA-N
isotopic purity   99.6 atom % D
assay   ≥99.00%
expl. lim.   0.34-6.3 % ((lit.))
packaging   pk of 10 × 1 mL
impurities   ≤0.0100% water
water
refractive index   n20/D 1.494 (lit.)
bp   110 °C(lit.)
mp   -84 °C (lit.)
density   0.943 g/mL at 25 °C (lit.)
mass shift   M+8
Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN1294 - class 3 - PG 2 - Toluene
WGK Germany 
2
Flash Point(F) 
39.2 °F
Flash Point(C) 
4 °C

Dispensette™ S
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Content

Pressure-Temperature Nomograph Interactive Tool

Use the interactive controls above to simplify calculations and improve the efficiency of your distillation or evaporation requirements.
Keywords: Evaporation, Vaporization

Properties of Solvents Table

The table below presents the physical characteristics of common solvents. Easily reference or compare solvent properties like boiling point, density, and refractive index all from one list.
Keywords: Gas chromatography, Green chemistry, High performance liquid chromatography, Melting, Solvents

Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?