• USA Home
  • 516414 - 1-(Phenylsulfonyl)-3-indolecarboxaldehyde

EMAIL THIS PAGE TO A FRIEND
516414 Sigma-Aldrich

1-(Phenylsulfonyl)-3-indolecarboxaldehyde

97%

Synonym: 1-(Benzenesulfonyl)indole-3-carboxaldehyde, 1-(Phenylsulfonyl)-3-formylindole, NSC 628191

  • CAS Number 80360-20-9

  • Empirical Formula (Hill Notation) C15H11NO3S

  • Molecular Weight 285.32

  •  MDL number MFCD02681985

  •  PubChem Substance ID 24873888

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C13 to C27, C13-C60, Carbonyl Compounds,
assay   97%
mp   157-161 °C (lit.)
storage temp.   room temp
SMILES string   [H]C(=O)c1cn(c2ccccc12)S(=O)(=O)c3ccccc3
InChI   1S/C15H11NO3S/c17-11-12-10-16(15-9-5-4-8-14(12)15)20(18,19)13-6-2-1-3-7-13/h1-11H
InChI key   ZLARITBCJILRBL-UHFFFAOYSA-N

Description

Application

Reactant for:
• Peterson olefination

Reactant for preparation of:
• Antifungal agents
• Potential topoisomerase II inhibitors
• Anti-inflammatory, analgesic and anticonvulsant agents
• Bacteriorhodopsin analogs
• Antitumor agents
• Antitubercular agents
• Inhibitors of human dUTPase and UNG2
• Uracil DNA glycosylase inhibitor
• Anti-HIV and anti-tumor β-carbolines

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Indoles

Sigma-Aldrich is proud to offer a new series of ChemFiles—called Privileged Structures, to our Drug Discovery and Organic Synthesis customers. Each piece will highlight a specific motif, selected app...
Chemfiles Volume 4 Article 8
Keywords: Building blocks, Cardiovascular, Diabetes, Diseases, Drug discovery, Medicinal chemistry, Organic synthesis, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?