• USA Home
  • 517100 - PSS-[3-(1,3-propanediol-2-ethyl-2-methyloxy]propyldimethylsilyloxy), heptacyclopentyl substituted

EMAIL THIS PAGE TO A FRIEND
517100 Sigma-Aldrich

PSS-[3-(1,3-propanediol-2-ethyl-2-methyloxy]propyldimethylsilyloxy), heptacyclopentyl substituted

Synonym: 2-Ethyl-2-[3-[(3,5,7,9,11,13,15-heptacyclopentylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxan-1-yloxy)dimethylsilyl]propoxymethyl]-1,3-propanediol, Diol-POSS®

  • CAS Number 268747-51-9

  • Empirical Formula (Hill Notation) C46H88O16Si9

  • Molecular Weight 1149.95

  •  MDL number MFCD02093702

Purchase

Properties

mp   285-290 °C (lit.)

Description

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?