• USA Home
  • 517518 - Scandium(III) bromide hydrate

EMAIL THIS PAGE TO A FRIEND
517518 Aldrich

Scandium(III) bromide hydrate

≥99.99%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Materials Science, Metal and Ceramic Science,
InChI Key   APPHYFNIXVIIJR-UHFFFAOYSA-K
assay   ≥99.99%
form   crystals and lumps
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?