• USA Home
  • 517623 - Dodecyl sulfate scandium(III) salt

EMAIL THIS PAGE TO A FRIEND
517623 Sigma-Aldrich

Dodecyl sulfate scandium(III) salt

Synonym: Scandium(III) dodecyl sulfate

Purchase

Properties

mp   159-169 °C (lit.)
SMILES string   [Sc+3].CCCCCCCCCCCCOS([O-])(=O)=O.CCCCCCCCCCCCOS([O-])(=O)=O.CCCCCCCCCCCCOS([O-])(=O)=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?