EMAIL THIS PAGE TO A FRIEND
518611 Sigma-Aldrich

Copper-zinc alloy

flakes, 1 mm

Synonym: Brass

Purchase

Properties

Related Categories Alloys, Materials Science, Metal and Ceramic Science More...
InChI Key   TVZPLCNGKSPOJA-UHFFFAOYSA-N
form   flakes
composition   copper, 50 wt. %
  zinc, 50 wt. %
particle size   1 mm
mp   ~870 °C (lit.)
density   1.17 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

520403 Copper-zinc alloy, powder, −60 mesh

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?