EMAIL THIS PAGE TO A FRIEND
519103 Sigma-Aldrich

Arsenic(V) sulfide

99.99%

Synonym: Arsenic pentasulfide, Diarsenic pentasulfide

Purchase

Properties

Related Categories Arsenic Catalysts, Catalysis and Inorganic Chemistry, Chalcogenides, Chemical Synthesis, Materials Science,
assay   99.99%
mp   >300 °C (lit.)
reaction suitability   reagent type: catalyst
SMILES string   S=[As](=S)S[As](=S)=S
InChI   1S/As2S5/c3-1(4)7-2(5)6
InChI key   AYRZLUSHOXJGKY-UHFFFAOYSA-N

Description

Application

• Used for preparation of glasses with small gold additions posessing photoluminescent and conductive properties
• Reactant for preparation of silver thioarsenate complex ethylenediammonium salt

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1557 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?