• USA Home
  • 519758 - Cartridge for Phosgene Generation

EMAIL THIS PAGE TO A FRIEND
519758 Sigma-Aldrich

Cartridge for Phosgene Generation

0.02 mole

Synonym: Triphosgene, Cartridge for Phosgene Generation

  • Empirical Formula (Hill Notation) C3Cl6O3

  • Molecular Weight 296.75

  •  EC Number 250-986-3

  •  MDL number MFCD00062848

  •  PubChem Substance ID 24873931

Purchase

Properties

Related Categories Chemical Synthesis, Coupling, Phosgene Derivatives, Synthetic Reagents
InChI Key   UCPYLLCMEDAXFR-UHFFFAOYSA-N
storage temp.   2-8°C

Description

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2928 8(6.1) / PGII
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Phosgene

Phosgene is an extremely versatile reagent allowing easy access to isocyanates, ureas, carbamates, carbonates, acyl and alkyl chlorides.1 Many of these can be used as reactive intermediates in peptid...
Matthias Junkers
ChemFiles 2007, 7.2, 4.
Keywords: Coupling reactions, Peptide synthesis

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?