• USA Home
  • 520756 - Palladium(II) oxide hydrate

EMAIL THIS PAGE TO A FRIEND
520756 Sigma-Aldrich

Palladium(II) oxide hydrate

≥99.9% trace metals basis

Synonym: Palladium monooxide hydrate, Palladium monoxide hydrate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Homogeneous Pd Catalysts, Materials Science, Metal and Ceramic Science,
InChI Key   CJCUJYURPLYQOL-UHFFFAOYSA-N
assay   ≥99.9% trace metals basis
form   powder
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?