EMAIL THIS PAGE TO A FRIEND
52208 Aldrich

1,15-Hexadecadiene

≥98.5% (GC)

  • CAS Number 21964-51-2

  • Linear Formula CH2=CH(CH2)12CH=CH2

  • Molecular Weight 222.41

  •  Beilstein Registry Number 1701162

  •  MDL number MFCD00216713

  •  PubChem Substance ID 24874248

Purchase

Properties

assay   ≥98.5% (GC)
refractive index   n20/D 1.447
bp   142-147 °C/6 mmHg(lit.)
mp   −14-−12 °C (lit.)
density   0.789 g/mL at 20 °C(lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?