• USA Home
  • 522333 - 1-[2-(Dimethylamino)ethyl]-1H-tetrazole-5-thiol

EMAIL THIS PAGE TO A FRIEND
522333 Sigma-Aldrich

1-[2-(Dimethylamino)ethyl]-1H-tetrazole-5-thiol

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Tetrazoles
Quality Level   100
assay   98%
availability   available only in USA
mp   215 °C (dec.) (lit.)
SMILES string   CN(C)CCn1nnnc1S
InChI   1S/C5H11N5S/c1-9(2)3-4-10-5(11)6-7-8-10/h3-4H2,1-2H3,(H,6,8,11)
InChI key   ODDAWJGQWOGBCX-UHFFFAOYSA-N

Description

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?