• USA Home
  • 522589 - Polycarbomethylsilane

EMAIL THIS PAGE TO A FRIEND
522589 Aldrich

Polycarbomethylsilane

average Mw ~800

Synonym: PCMS, Poly[(methylsilylene)methylene], Polycarbosilane

Purchase

Description

Application

Polymeric precursor of SiC ceramics. Ceramic coating from polymer melt or solution.

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

Electrospinning: An Enabling Technique for Nanostructured Materials

Fibrous nanomaterials are attractive for a range of applications due to their intrinsically high porosities and large surface areas. Electrospinning is a simple, versatile technique for generating na...
Dr. Jingwei Xie and Prof. Younan Xia
Material Matters 2008, 3.1, 19.
Keywords: Applications, Biomaterials, Capabilities, Cell biology, Central Nervous System, Ceramics, Compliance, Evaporation, Growth factors, Methods, Nanomaterials, Nanotechnology, Nanotubes, Separation, Sol-gel, Stem cell biology, Tools, Type

Micro and Nanometer Scale Photopatterning of Self-Assembled Monolayers

Graham J. Leggett* Department of Chemistry, University of Sheffield, Brook Hill, Sheffield S3 7HF, United Kingdom *Email: Graham.Leggett@shef.ac.uk
Keywords: Absorption, Adsorption, Degradations, Deposition, Microscopy, Optical microscopy, Oxidations, Polymerization reactions, Semiconductor, Spectroscopy

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?