• USA Home
  • 523593 - 1,1,3,3,5,5-Hexamethyl-1,5-bis[2-(5-norbornen-2-yl)ethyl]trisiloxane

EMAIL THIS PAGE TO A FRIEND
523593 Sigma-Aldrich

1,1,3,3,5,5-Hexamethyl-1,5-bis[2-(5-norbornen-2-yl)ethyl]trisiloxane

mixture of endo and exo

Purchase

Properties

Related Categories Chemical Synthesis, Materials Science, Monomers, Nanomaterials, Organometallic Reagents,
refractive index   n20/D 1.4690 (lit.)
density   0.976 g/mL at 25 °C (lit.)
SMILES string   C[Si](C)(CCC1CC2CC1C=C2)O[Si](C)(C)O[Si](C)(C)CCC3CC4CC3C=C4
InChI   1S/C24H44O2Si3/c1-27(2,13-11-23-17-19-7-9-21(23)15-19)25-29(5,6)26-28(3,4)14-12-24-18-20-8-10-22(24)16-20/h7-10,19-24H,11-18H2,1-6H3
InChI key   OJLPCKNKLYMMBA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Etch-Resistant Block Copolymers

Block copolymers offer a means of combining the desirable characteristics of different polymers in a new hybrid material. Polymers consisting of hydrophobic and hydrophilic blocks, for example, can b...
Prof. Padma GopalanProf. Shu Yang
Material Matters 2006, 1.1, 6.
Keywords: Applications, Material Matters, Materials Science, Microscopy, Ozonolysis, Polymerization reactions, Purification, Radical polymerization, Reversible addition-fragmentation chain transfer polymerizations, Scanning electron microscopy, Semiconductor, Transmission electron microscopy

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?