• USA Home
  • 523658 - endo-3,7,14-Tris(dimethylvinylsilyloxy)-1,3,5,7,9,11,14-heptacyclopentyltricyclo[7.3.3.15,11]heptasiloxane

EMAIL THIS PAGE TO A FRIEND
523658 Sigma-Aldrich

endo-3,7,14-Tris(dimethylvinylsilyloxy)-1,3,5,7,9,11,14-heptacyclopentyltricyclo[7.3.3.15,11]heptasiloxane

Synonym: Tris(dimethylvinylsilyloxy)-POSS®

  • CAS Number 312693-39-3

  • Empirical Formula (Hill Notation) C47H90O12Si10

  • Molecular Weight 1128.07

  •  MDL number MFCD02093773

  •  PubChem Substance ID 24874354

Purchase

Properties

mp   >350 °C (lit.)
SMILES string   C[Si](C)(O[Si]1(O[Si]2(O[Si](O[Si](C)(C)C=C)(O[Si]3(O[Si](O[Si](C)(C)C=C)(O[Si](O1)(O[Si](O2)(O3)C4CCCC4)C5CCCC5)C6CCCC6)C7CCCC7)C8CCCC8)C9CCCC9)C%10CCCC%10)C=C

Description

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?