• USA Home
  • 524166 - (Propylcyclopentadienyl)molybdenum(I) tricarbonyl dimer

EMAIL THIS PAGE TO A FRIEND
524166 Sigma-Aldrich

(Propylcyclopentadienyl)molybdenum(I) tricarbonyl dimer

Synonym: 1,1,3,3-Tetramethyl-1,3-bis[2-(5-norbornen-2-yl)ethyl]disiloxane

Purchase

Properties

Related Categories CVD and ALD Precursors by Metal, Materials Science, Micro/NanoElectronics, Molybdenum, Vapor Deposition Precursors More...
form   solid
mp   152 °C (dec.) (lit.)
SMILES string   [Mo][Mo].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].CCC[C]1[CH][CH][CH][CH]1.CCC[C]2[CH][CH][CH][CH]2
InChI   1S/2C8H11.6CO.2Mo/c2*1-2-5-8-6-3-4-7-8;6*1-2;;/h2*3-4,6-7H,2,5H2,1H3;;;;;;;;
InChI key   KKIIOHGMKWAYPK-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?