• USA Home
  • 525030 - 2-Ethylbenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
525030 Sigma-Aldrich

2-Ethylbenzyl alcohol

98%

  • CAS Number 767-90-8

  • Linear Formula C2H5C6H4CH2OH

  • Molecular Weight 136.19

  •  MDL number MFCD03093889

  •  PubChem Substance ID 24874463

  •  NACRES NA.22

Purchase

Properties

Related Categories Alcohols, Building Blocks, C9 to C10, Chemical Synthesis, Organic Building Blocks,
assay   98%
refractive index   n20/D 1.536 (lit.)
bp   229 °C (lit.)
density   1.011 g/mL at 25 °C (lit.)
SMILES string   CCc1ccccc1CO
InChI   1S/C9H12O/c1-2-8-5-3-4-6-9(8)7-10/h3-6,10H,2,7H2,1H3
InChI key   SBUIQTMDIOLKAL-UHFFFAOYSA-N

Description

General description

2-Ethylbenzyl alcohol is a benzyl alcohol derivative. Kinetic studies show that the reaction is first-order in both alcohol and oxidant" and forms"corresponding benzaldehydes in acetonitrile and water.

Application

2-Ethylbenzyl alcohol may be used to synthesize 2-ethylbenzyl chloride.

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
235.4 °F - closed cup
Flash Point(C) 
113 °C - closed cup

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?