• USA Home
  • 525545 - (S)-(−)-N-Allyl-α-methylbenzylamine

EMAIL THIS PAGE TO A FRIEND
525545 Sigma-Aldrich

(S)-(−)-N-Allyl-α-methylbenzylamine

97%

Purchase

Properties

assay   97%
optical activity   [α]20/D −56°, c = 1 in methylene chloride
refractive index   n20/D 1.5145 (lit.)
bp   58-59 °C/1 mmHg (lit.)
density   0.919 g/mL at 25 °C (lit.)
SMILES string   C[C@H](NCC=C)c1ccccc1
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1760 8 / PGIII
WGK Germany 
3
Flash Point(F) 
186.8 °F
Flash Point(C) 
86 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?