• USA Home
  • 52647 - Hexamethylenediamine-N,N,N′,N′-tetrakis(methylphosphonic acid) solution

EMAIL THIS PAGE TO A FRIEND
52647 Sigma-Aldrich

Hexamethylenediamine-N,N,N′,N′-tetrakis(methylphosphonic acid) solution

technical, ~25% in H2O (T)

Synonym: Dequest® 2054

  • CAS Number 23605-74-5

  • Empirical Formula (Hill Notation) C10H28N2O12P4

  • Molecular Weight 492.23

  •  Beilstein/REAXYS Number 2405158

  •  EC Number 231-791-2

  •  MDL number MFCD00058890

  •  PubChem Substance ID 329757866

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Phosphonic and Phosphinic Acids, Phosphorus Compounds More...
InChI Key   KIDJHPQACZGFTI-UHFFFAOYSA-N
grade   technical
concentration   ~25% in H2O (T)

Description

Legal Information

Dequest is a registered trademark of Dequest AG

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?