• USA Home
  • 527297 - PSS-Methylphenylvinylsilyloxy-Heptacyclopentyl substituted

EMAIL THIS PAGE TO A FRIEND
527297 Sigma-Aldrich

PSS-Methylphenylvinylsilyloxy-Heptacyclopentyl substituted

97%

Synonym: (Methylphenylvinylsilyloxy)heptacyclopentylpentacyclooctasiloxane, Methylphenylvinylsilyloxy-POSS®

  • CAS Number 312693-48-4

  • Empirical Formula (Hill Notation) C44H74O13Si9

  • Molecular Weight 1063.82

  •  MDL number MFCD02683495

Purchase

Properties

assay   97%
mp   245-249 °C (lit.)

Description

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?