EMAIL THIS PAGE TO A FRIEND
528048 Sigma-Aldrich

1-Octanethiol

≥98.5%

Synonym: n-Octyl mercaptan, Mercaptan C8, NOM, Octyl mercaptan

  • CAS Number 111-88-6

  • Linear Formula CH3(CH2)7SH

  • Molecular Weight 146.29

  •  Beilstein/REAXYS Number 1733101

  •  EC Number 203-918-1

  •  MDL number MFCD00004912

Purchase

Properties

assay   ≥98.5%
availability   available only in EU
refractive index   n20/D 1.452 (lit.)
bp   197-200 °C (lit.)
density   0.843 g/mL at 25 °C (lit.)
SMILES string   CCCCCCCCS
InChI   1S/C8H18S/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3
InChI key   KZCOBXFFBQJQHH-UHFFFAOYSA-N

Description

Legal Information

Product of Arkema

NOM is a trademark of Elf Atochem

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
WGK 3
Flash Point(F) 
158.0 °F
Flash Point(C) 
70 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

471836 1-Octanethiol, ≥98.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?