• USA Home
  • 528447 - Ethyl 2-ethoxycinnamate

EMAIL THIS PAGE TO A FRIEND
528447 Sigma-Aldrich

Ethyl 2-ethoxycinnamate

97%

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
assay   97%
refractive index   n20/D 1.56 (lit.)
bp   310 °C (lit.)
density   1.06 g/mL at 25 °C (lit.)
SMILES string   CCOC(=O)\C=C\c1ccccc1OCC
InChI   1S/C13H16O3/c1-3-15-12-8-6-5-7-11(12)9-10-13(14)16-4-2/h5-10H,3-4H2,1-2H3/b10-9+
InChI key   WJRLRVSUBGIUGI-MDZDMXLPSA-N

Description

Packaging

5 mL in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?