• USA Home
  • 529052 - 2,5-Diacetoxytoluene

EMAIL THIS PAGE TO A FRIEND
529052 Sigma-Aldrich

2,5-Diacetoxytoluene

97%

Purchase

Properties

Related Categories Building Blocks, C10 to C11, Carbonyl Compounds, Chemical Synthesis, Esters,
assay   97%
mp   41-45 °C (lit.)
storage temp.   room temp
SMILES string   CC(=O)Oc1ccc(OC(C)=O)c(C)c1
InChI   1S/C11H12O4/c1-7-6-10(14-8(2)12)4-5-11(7)15-9(3)13/h4-6H,1-3H3
InChI key   KUZVIVNLNXNLAQ-UHFFFAOYSA-N

Description

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?